Learn More
Bachem g2-MSH
SDP
Supplier:  Bachem 4008709.0001
Encompass_Preferred
g2-MSH , H-Tyr-Val-Met-Gly-His-Phe-Arg-Trp-Asp-Arg-Phe-Gly-OH, 72711-43-4, C74H99N21O16S, Mr 1570.80, 1mg. g-MSH. g2-MSH has been located in the cryptic region of the ACTH/b-lipotropin precursor protein from the intermediate lobe of bovine pituitary.
Catalog No. 50-259-841