Learn More
Grainger G2228 6-Inch Work Boot M 9 Brown PR
Catalog No. 502765095
Encompass_Preferred
The Carhartt 6 Inch Work Boot is a rugged leather work boot designed for durability and comfort. It features an oil-tanned leather upper, Ortholite insole for cushioning, Rugged Flex technology for flexibility, and a lightweight EVA midsole. The boot has a padded collar and tongue for added comfort. It has a cement construction which is directly attached to the outsole for flexibility and reduced break-in time. The outsole is made of durable rubber lug for traction. This work boot meets ASTM F2413-18 M I/75 C/75 standards for impact and compression resistance. It comes in brown and black leather.
Catalog No. 50-276-5095 Supplier Grainger Supplier No. 16P470
The Carhartt 6 Inch Work Boot is a rugged leather work boot designed for durability and comfort. It features an oil-tanned leather upper, Ortholite insole for cushioning, Rugged Flex technology for flexibility, and a lightweight EVA midsole. The boot has a padded collar and tongue for added comfort. It has a cement construction which is directly attached to the outsole for flexibility and reduced break-in time. The outsole is made of durable rubber lug for traction. This work boot meets ASTM F2413-18 M I/75 C/75 standards for impact and compression resistance. It comes in brown and black leather.