Learn More
Grainger G2228 6-Inch Work Boot W 11 1/2 Brown PR
Catalog No. 502768374
Encompass_Preferred
The Grainger 16P481 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon and synthetic fibers. This 8" x 4" hand pad has a medium duty scrubbing level and is ideal for cleaning pots, pans, dishes, flatware, counters, walls, appliances, tools, equipment, and more. It effectively removes grease, dirt, food residue, and other grime without scratching. The pad has an attached loop for easy hanging and storage. It provides economical scrubbing power.
Catalog No. 50-276-8374 Supplier Grainger Supplier No. 16P481
The Grainger 16P481 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon and synthetic fibers. This 8" x 4" hand pad has a medium duty scrubbing level and is ideal for cleaning pots, pans, dishes, flatware, counters, walls, appliances, tools, equipment, and more. It effectively removes grease, dirt, food residue, and other grime without scratching. The pad has an attached loop for easy hanging and storage. It provides economical scrubbing power.