Learn More
Grainger G2228 6-Inch Work Boot W 12 Brown PR
Catalog No. 502768268
Encompass_Preferred
The Grainger G2228 6-Inch Work Boot is a brown leather work boot with a lace-up design and a steel safety toe. It has an oil-resistant outsole and a removable cushion insole for comfort. The boot meets ASTM F2413-18 I/75 C/75 standards and has electrical hazard protection. It comes in sizes 7-15, including half sizes, and has a 6 inch height. The boot is designed for general work environments.
Catalog No. 50-276-8268 Supplier Grainger Supplier No. 16P483
The Grainger G2228 6-Inch Work Boot is a brown leather work boot with a lace-up design and a steel safety toe. It has an oil-resistant outsole and a removable cushion insole for comfort. The boot meets ASTM F2413-18 I/75 C/75 standards and has electrical hazard protection. It comes in sizes 7-15, including half sizes, and has a 6 inch height. The boot is designed for general work environments.