Learn More
Grainger G2228 6-Inch Work Boot W 8 Brown PR
Catalog No. 502768404
Encompass_Preferred
The Grainger G2228 6-Inch Work Boot is a brown leather work boot with a steel toe and oil-resistant outsole. It has a lace-up design with speed hooks for easy on and off, a padded collar and tongue for comfort, and a removable cushion insole. The boot meets ASTM F2413-18 I/75 C/75 standards and has an electrical hazard rating. It is designed for general work environments.
Catalog No. 50-276-8404 Supplier Grainger Supplier No. 16P467
The Grainger G2228 6-Inch Work Boot is a brown leather work boot with a steel toe and oil-resistant outsole. It has a lace-up design with speed hooks for easy on and off, a padded collar and tongue for comfort, and a removable cushion insole. The boot meets ASTM F2413-18 I/75 C/75 standards and has an electrical hazard rating. It is designed for general work environments.