Learn More
Grainger G2233 Western Boot M 13 Brown PR
Catalog No. 502765020
Encompass_Preferred
The Grainger 16P773 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is thermally protected and rated for continuous duty applications. It can provide 0.56 kW of output power. The 16P773 has a NEMA standard 56C frame size with standard starting torque and breakdown torque. This electric motor is suitable for various industrial applications requiring up to 3/4 horsepower.
Catalog No. 50-276-5020 Supplier Grainger Supplier No. 16P773
The Grainger 16P773 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is thermally protected and rated for continuous duty applications. It can provide 0.56 kW of output power. The 16P773 has a NEMA standard 56C frame size with standard starting torque and breakdown torque. This electric motor is suitable for various industrial applications requiring up to 3/4 horsepower.