Learn More
Grainger G2233 Western Boot M 8 1/2 Brown PR
Catalog No. 502767739
Encompass_Preferred
The Carhartt Men's 11" Waterproof Insulated Western Boot is a rugged, durable boot designed for work in tough outdoor conditions. It features a full-grain leather and Cordura nylon upper with a waterproof membrane to keep feet dry. The boot has 200g 3M Thinsulate Ultra insulation for warmth in cold weather. It has an oil-tanned leather vamp and shaft overlay for abrasion resistance. The outsole is rugged rubber lug with a 1.5" heel. The boot opening measures approximately 14.5" around. Other features include an Ortholite insole for cushioning and odor control and triple-stitched seams for durability. This is an ideal boot for working outdoors in wet, cold conditions.
Catalog No. 50-276-7739 Supplier Grainger Supplier No. 16P757
The Carhartt Men's 11" Waterproof Insulated Western Boot is a rugged, durable boot designed for work in tough outdoor conditions. It features a full-grain leather and Cordura nylon upper with a waterproof membrane to keep feet dry. The boot has 200g 3M Thinsulate Ultra insulation for warmth in cold weather. It has an oil-tanned leather vamp and shaft overlay for abrasion resistance. The outsole is rugged rubber lug with a 1.5" heel. The boot opening measures approximately 14.5" around. Other features include an Ortholite insole for cushioning and odor control and triple-stitched seams for durability. This is an ideal boot for working outdoors in wet, cold conditions.