Learn More
Aviva Systems Biology Corp G22P1 Antibody - N-terminal region (ARP34117_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34117_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against G22P1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human G22P1. Alternate names: ML8, KU70, TLAA, CTC75, CTCBF, G22P1ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55255