Learn More
Grainger G2350 Webbing Nylon 3/4 W Orange
Catalog No. 502750899
Encompass_Preferred
The Bulk Strap Bulk Webbing Webbing is a 1-inch wide polypropylene webbing strap sold in a 500 foot roll. It has a breaking strength of 1,200 lbs and is UV treated for outdoor use. The strap is blue in color. It can be used for a variety of purposes including bundling, securing cargo, and craft projects. The strap meets military specification MIL-W-4088 Type III and is made in the USA.
Catalog No. 50-275-0899 Supplier Grainger Supplier No. 14A806
The Bulk Strap Bulk Webbing Webbing is a 1-inch wide polypropylene webbing strap sold in a 500 foot roll. It has a breaking strength of 1,200 lbs and is UV treated for outdoor use. The strap is blue in color. It can be used for a variety of purposes including bundling, securing cargo, and craft projects. The strap meets military specification MIL-W-4088 Type III and is made in the USA.