Learn More
Grainger BULK-STRAP Webbing, Nylon, 3,800 lb Tensile Strength, 1,250 lb Working Load Limit, 1 in x 102 ft, 1 ea
Catalog No. 502750665
Encompass_Preferred
This medium-duty nylon webbing is engineered for versatile use in load securing and cargo control applications. Constructed from durable nylon, the material features high abrasion resistance and consistent strength for reliable performance in various industrial and commercial environments. Please note that this product is not intended for lifting applications.
  • Tensile strength of 3,800 lb
  • Working load limit of 1,250 lb
  • Dimensions of 102 ft length by 1 in width
  • Thickness of 1/16 in
  • Orange color for high visibility
  • Suitable for applications including tie-down straps, trucking, banners, luggage, and recreational equipment
Catalog No. 50-275-0665 Supplier Grainger Supplier No. 14A812
This medium-duty nylon webbing is engineered for versatile use in load securing and cargo control applications. Constructed from durable nylon, the material features high abrasion resistance and consistent strength for reliable performance in various industrial and commercial environments. Please note that this product is not intended for lifting applications.
  • Tensile strength of 3,800 lb
  • Working load limit of 1,250 lb
  • Dimensions of 102 ft length by 1 in width
  • Thickness of 1/16 in
  • Orange color for high visibility
  • Suitable for applications including tie-down straps, trucking, banners, luggage, and recreational equipment