Learn More
Grainger BULK-STRAP Webbing, Nylon, 7,000 lb/in Tensile Strength, 27 ft, 1 ea
Catalog No. 502750771
Encompass_Preferred
This medium-duty nylon webbing is designed for general load securing and cargo control applications. It features high tensile strength and provides good elasticity and recovery under tension. The material is abrasion resistant and suitable for a variety of industrial, commercial, and recreational uses.
  • Tensile strength of 7,000 lb per inch
  • Working load limit of 2,300 lb
  • 27 ft length
  • Abrasion, UV, and mildew resistant
  • Absorbs moisture and dries quickly
  • Suitable for applications including trucking, cargo control, construction, and marine equipment
Catalog No. 50-275-0771 Supplier Grainger Supplier No. 14A789
This medium-duty nylon webbing is designed for general load securing and cargo control applications. It features high tensile strength and provides good elasticity and recovery under tension. The material is abrasion resistant and suitable for a variety of industrial, commercial, and recreational uses.
  • Tensile strength of 7,000 lb per inch
  • Working load limit of 2,300 lb
  • 27 ft length
  • Abrasion, UV, and mildew resistant
  • Absorbs moisture and dries quickly
  • Suitable for applications including trucking, cargo control, construction, and marine equipment