Learn More
Grainger G2363 Webbing Nylon 2 W Blue
Catalog No. 502750693
Encompass_Preferred
The Bulk Strap Bulk Webbing Webbing is a 1-inch wide polypropylene webbing strap sold in a 500 foot roll. It is lightweight and flexible with a breaking strength of 1,200 lbs. The strap is UV treated for outdoor use and has heat sealed edges to prevent fraying. It can be used for a variety of purposes including tie downs, tow straps, slings, cargo nets and more. The strap meets military specification MIL-W-4088 Type III and is made in the USA.
Catalog No. 50-275-0693 Supplier Grainger Supplier No. 14A790
The Bulk Strap Bulk Webbing Webbing is a 1-inch wide polypropylene webbing strap sold in a 500 foot roll. It is lightweight and flexible with a breaking strength of 1,200 lbs. The strap is UV treated for outdoor use and has heat sealed edges to prevent fraying. It can be used for a variety of purposes including tie downs, tow straps, slings, cargo nets and more. The strap meets military specification MIL-W-4088 Type III and is made in the USA.