Learn More
Grainger G2373 Webbing Polypropylene 1 1/2 W Orange
Catalog No. 502750744
Encompass_Preferred
The Grainger 14A829 is a 14 gauge, type 1 electrical extension cord that is 25 feet long. It has a yellow jacket and is designed for indoor and outdoor use. The 3-wire grounded cord has a molded plug on one end and receptacle on the other end. It can handle up to 15 amps and 125 volts. The flexible vinyl jacket resists oil, water and abrasion. This general purpose extension cord is UL listed and CSA certified.
Catalog No. 50-275-0744 Supplier Grainger Supplier No. 14A829
The Grainger 14A829 is a 14 gauge, type 1 electrical extension cord that is 25 feet long. It has a yellow jacket and is designed for indoor and outdoor use. The 3-wire grounded cord has a molded plug on one end and receptacle on the other end. It can handle up to 15 amps and 125 volts. The flexible vinyl jacket resists oil, water and abrasion. This general purpose extension cord is UL listed and CSA certified.