Learn More
Grainger G2374 Webbing Polypropylene 1 1/2 W Orange
Catalog No. 502751022
Encompass_Preferred
The Grainger 14A830 is a Westward digital multimeter that measures AC/DC voltage, DC current, resistance, diode test, and continuity. It has a large LCD display with 0.75" digits, manual ranging for increased accuracy, protective holster with probe holders, and includes test leads. This general purpose multimeter is compact, portable, and easy to use for basic electrical testing and troubleshooting.
Catalog No. 50-275-1022 Supplier Grainger Supplier No. 14A830
The Grainger 14A830 is a Westward digital multimeter that measures AC/DC voltage, DC current, resistance, diode test, and continuity. It has a large LCD display with 0.75" digits, manual ranging for increased accuracy, protective holster with probe holders, and includes test leads. This general purpose multimeter is compact, portable, and easy to use for basic electrical testing and troubleshooting.