Learn More
Grainger G2375 Webbing Polypropylene 2 W Orange
Catalog No. 502750801
Encompass_Preferred
The Grainger Approved 2" Wide Orange Polypropylene Webbing is a strong, lightweight webbing made of polypropylene and measuring 2" wide. It features excellent resistance to rot, mildew and abrasion. This orange webbing has a breaking strength of 1,200 lbs and meets military specification MIL-W-4088 Type VIII for quality assurance. It can be used for a variety of applications including tie downs, tow straps, slings, harnesses and more. This polypropylene webbing comes in a 100 ft roll and is an ideal choice for industrial, commercial and DIY projects where high visibility orange webbing is needed.
Catalog No. 50-275-0801 Supplier Grainger Supplier No. 14A831
The Grainger Approved 2" Wide Orange Polypropylene Webbing is a strong, lightweight webbing made of polypropylene and measuring 2" wide. It features excellent resistance to rot, mildew and abrasion. This orange webbing has a breaking strength of 1,200 lbs and meets military specification MIL-W-4088 Type VIII for quality assurance. It can be used for a variety of applications including tie downs, tow straps, slings, harnesses and more. This polypropylene webbing comes in a 100 ft roll and is an ideal choice for industrial, commercial and DIY projects where high visibility orange webbing is needed.