Learn More
Grainger G2386 Arc Flash Body Harness ExoFit NEX XL
Catalog No. 502766671
Encompass_Preferred
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for fall protection when working at height. It is made with flame resistant webbing that meets NFPA 70E arc flash protection requirements. The harness has a back D-ring for fall arrest and shoulder, chest, and leg D-rings for positioning. It comes in sizes ranging from small to XX-large to provide a customized fit. The harness is ANSI and OSHA compliant and meets applicable ASTM standards. It provides a safe, comfortable option for those working at height in environments with arc flash hazards.
Catalog No. 50-276-6671 Supplier Grainger Supplier No. 16V459
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for fall protection when working at height. It is made with flame resistant webbing that meets NFPA 70E arc flash protection requirements. The harness has a back D-ring for fall arrest and shoulder, chest, and leg D-rings for positioning. It comes in sizes ranging from small to XX-large to provide a customized fit. The harness is ANSI and OSHA compliant and meets applicable ASTM standards. It provides a safe, comfortable option for those working at height in environments with arc flash hazards.