Learn More
Grainger G2386 Arc Flash Body Harness ExoFit NEX M
Catalog No. 502766685
Encompass_Preferred
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for arc flash protection. It is made with flame resistant webbing and has an arc rating of 40 cal/cm2. The harness has a back D-ring for fall arrest and features sub-pelvic support straps for comfort when suspended. It comes in sizes ranging from small to 4XL. This harness meets applicable OSHA, ANSI and CSA standards. It provides arc flash protection along with fall protection.
Catalog No. 50-276-6685 Supplier Grainger Supplier No. 16V457
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for arc flash protection. It is made with flame resistant webbing and has an arc rating of 40 cal/cm2. The harness has a back D-ring for fall arrest and features sub-pelvic support straps for comfort when suspended. It comes in sizes ranging from small to 4XL. This harness meets applicable OSHA, ANSI and CSA standards. It provides arc flash protection along with fall protection.