Learn More
Grainger G2387 Arc Flash Body Harness ExoFit NEX L
Catalog No. 502765359
Encompass_Preferred
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for fall protection and arc flash protection. It is made with flame resistant webbing and has a back D-ring for fall arrest. The harness has a quick connect chest strap, leg straps, and sub-pelvic strap for comfort and security. It meets ASTM F887 standards and comes in sizes ranging from small to 4XL. The arc flash protection helps provide a path to ground in the event of an arc flash.
Catalog No. 50-276-5359 Supplier Grainger Supplier No. 16V462
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for fall protection and arc flash protection. It is made with flame resistant webbing and has a back D-ring for fall arrest. The harness has a quick connect chest strap, leg straps, and sub-pelvic strap for comfort and security. It meets ASTM F887 standards and comes in sizes ranging from small to 4XL. The arc flash protection helps provide a path to ground in the event of an arc flash.