Learn More
Electron Microscopy Sciences Walton & Beckett Graticule G25, 16 mm,
SDP
Catalog No. 5029682
Encompass_Preferred
The Walton and Beckett graticule is for counting fibrous dust (asbestos or glass fibers) and particularly where the majority of fibers to be counted are shorter than 5 microns. The circle is divided into four by two diametrical lines scaled in units of 5 and 3 microns respectively. 3 and 5 microns are the critical measurements of fiber lengths and diameters used in fiber counting. Walton and Beckett include shapes with an aspect ratio of 3:1 or 5:1 for comparing to fibers.
IMPORTANT NOTE: The circle on these Walton & Beckett graticules must represent 100 microns at the stage and each one must be manufactured to the individual instrument. Based on the G22. The G25 is produced by the Institute of Occupational Health.
Size: 16 mm
Catalog No. 50-296-82 Supplier Electron Microscopy Sciences Supplier No. 6803016
The Walton and Beckett graticule is for counting fibrous dust (asbestos or glass fibers) and particularly where the majority of fibers to be counted are shorter than 5 microns. The circle is divided into four by two diametrical lines scaled in units of 5 and 3 microns respectively. 3 and 5 microns are the critical measurements of fiber lengths and diameters used in fiber counting. Walton and Beckett include shapes with an aspect ratio of 3:1 or 5:1 for comparing to fibers.
IMPORTANT NOTE: The circle on these Walton & Beckett graticules must represent 100 microns at the stage and each one must be manufactured to the individual instrument. Based on the G22. The G25 is produced by the Institute of Occupational Health.
Size: 16 mm