Learn More
Medchemexpress LLC G3-C12 TFA | 99.4% | 1873.00 | 5 MG
SDP
Catalog No. 5000424299
Encompass_Preferred
G3-C12 (TFA) is a galectin-3 binding peptide that exhibits anticancer activity with a Kd of 88 nM. It shows high affinity for galectin-3 but no affinity for other galectin family members or other lectins. When bearing opolymers N-(2-hydroxypropyl)methacrylamide (HPMA), it potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3, demonstrating superior targetability to galectin-3 compared to galactose bearing copolymer.
  • Galectin-3 binding peptide
  • Kd of 88 nM
  • Shows anticancer activity
  • High affinity for galectin-3
  • No affinity for other galectin family members or lectins
  • Potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3 when conjugated with HPMA opolymers
Catalog No. 50-004-24299 Supplier Medchemexpress LLC Supplier No. HYP1592A5MG
G3-C12 (TFA) is a galectin-3 binding peptide that exhibits anticancer activity with a Kd of 88 nM. It shows high affinity for galectin-3 but no affinity for other galectin family members or other lectins. When bearing opolymers N-(2-hydroxypropyl)methacrylamide (HPMA), it potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3, demonstrating superior targetability to galectin-3 compared to galactose bearing copolymer.
  • Galectin-3 binding peptide
  • Kd of 88 nM
  • Shows anticancer activity
  • High affinity for galectin-3
  • No affinity for other galectin family members or lectins
  • Potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3 when conjugated with HPMA opolymers