Learn More
Invivogen G3-YSD Control, Control for cGAS Agonist G3-YSD, 200 µg
SDP
Catalog No. NC1665844
Encompass
G3-YSD Control is a negative control for G3-YSD, a potent cGAS (cyclic GMP-AMP synthase, cGAMP synthase) agonist. This negative control differs from the G3-YSD agonist in the hairpin-flanking nucleoside trimers.
Catalog No. NC1665844 Supplier Invivogen Supplier No. TLRLYDNAC
G3-YSD Control is a negative control for G3-YSD, a potent cGAS (cyclic GMP-AMP synthase, cGAMP synthase) agonist. This negative control differs from the G3-YSD agonist in the hairpin-flanking nucleoside trimers.