Learn More
Grainger G3254 Therma Level Gauge HDPE D 23in
Catalog No. 502765324
Encompass_Preferred
The Grainger Approved Therma Level Gauge HDPE is a high-density polyethylene level gauge designed for use with water heaters and boilers. It features a clear, shatter-resistant tube with easy-to-read level markings and a 3/4" NPT fitting. The gauge allows visual monitoring of system water levels and can withstand temperatures up to 200°F. With dimensions of 18" length x 2" width, this durable HDPE construction level gauge provides an economical solution for monitoring water levels in hydronic heating systems.
Catalog No. 50-276-5324 Supplier Grainger Supplier No. 16N618
The Grainger Approved Therma Level Gauge HDPE is a high-density polyethylene level gauge designed for use with water heaters and boilers. It features a clear, shatter-resistant tube with easy-to-read level markings and a 3/4" NPT fitting. The gauge allows visual monitoring of system water levels and can withstand temperatures up to 200°F. With dimensions of 18" length x 2" width, this durable HDPE construction level gauge provides an economical solution for monitoring water levels in hydronic heating systems.