Learn More
Grainger G3255 Therma Level Gauge HDPE D 57in
Catalog No. 502768424
Encompass_Preferred
The Grainger Approved Therma Level Gauge is a liquid-filled thermometer designed for use in industrial boilers, water heaters, and process tanks to indicate liquid levels. This stainless steel gauge has a 10 inch scale graduated from 30 to 240 degrees Fahrenheit. The red liquid inside expands with increasing temperature and contracts with decreasing temperature to accurately display the temperature inside the tank. This allows operators to monitor tank conditions and fluid levels. The gauge has 1/2 inch NPT connections and is made in the USA.
Catalog No. 50-276-8424 Supplier Grainger Supplier No. 16N603
The Grainger Approved Therma Level Gauge is a liquid-filled thermometer designed for use in industrial boilers, water heaters, and process tanks to indicate liquid levels. This stainless steel gauge has a 10 inch scale graduated from 30 to 240 degrees Fahrenheit. The red liquid inside expands with increasing temperature and contracts with decreasing temperature to accurately display the temperature inside the tank. This allows operators to monitor tank conditions and fluid levels. The gauge has 1/2 inch NPT connections and is made in the USA.