Learn More
Grainger G3256 Therma Level Gauge HDPE D 57in
Catalog No. 502766913
Encompass_Preferred
The Grainger 16N604 is a 16 oz. spray bottle made of HDPE plastic with a 28/400 neck finish. It has a trigger sprayer top and can hold liquids. The bottle is translucent and measures 8-1/2" tall x 2-3/4" diameter. It can be used for spraying cleaning chemicals, automotive chemicals, pesticides, herbicides, and other general industrial liquids. The working pressure is 30 PSI with a maximum temperature resistance of 120°F. This durable and reusable bottle is sold individually.
Catalog No. 50-276-6913 Supplier Grainger Supplier No. 16N604
The Grainger 16N604 is a 16 oz. spray bottle made of HDPE plastic with a 28/400 neck finish. It has a trigger sprayer top and can hold liquids. The bottle is translucent and measures 8-1/2" tall x 2-3/4" diameter. It can be used for spraying cleaning chemicals, automotive chemicals, pesticides, herbicides, and other general industrial liquids. The working pressure is 30 PSI with a maximum temperature resistance of 120°F. This durable and reusable bottle is sold individually.