Learn More
Grainger G3257 Therma Level Gauge HDPE D 36in
Catalog No. 502767108
Encompass_Preferred
The Grainger Approved Therma Level Gauge HDPE is a high-density polyethylene level gauge designed for use with water heaters and boilers. It features a clear, shatter-resistant tube with easy-to-read level markings and temperature ratings up to 180°F. This level gauge has 3/4 in. NPT male thread connections and measures 16 in. long, allowing it to be installed on tanks up to 14 in. deep. The heavy-duty plastic construction provides chemical resistance and durability. Overall, this is a durable, easy-to-install level gauge suitable for monitoring water levels in residential and commercial water heating applications.
Catalog No. 50-276-7108 Supplier Grainger Supplier No. 16N624
The Grainger Approved Therma Level Gauge HDPE is a high-density polyethylene level gauge designed for use with water heaters and boilers. It features a clear, shatter-resistant tube with easy-to-read level markings and temperature ratings up to 180°F. This level gauge has 3/4 in. NPT male thread connections and measures 16 in. long, allowing it to be installed on tanks up to 14 in. deep. The heavy-duty plastic construction provides chemical resistance and durability. Overall, this is a durable, easy-to-install level gauge suitable for monitoring water levels in residential and commercial water heating applications.