Learn More
Grainger G3258 Therma Level Gauge HDPE D 46in
Catalog No. 502764863
Encompass_Preferred
The Grainger 16N602 is a 1/4 HP, 115V, 60Hz, 1-phase, 3450 RPM, 56C frame, open drip proof, ball bearing, C-face, sleeve bearing electric motor. It has a NEMA 48 frame size with a 1.25" shaft diameter and 1.38" shaft height. The motor is totally enclosed and fan cooled with a Class B insulation and 1.15 service factor. It can output 0.186 kW with a full load amp rating of 5.8 Amps. The 16N602 electric motor is ideal for various light industrial applications.
Catalog No. 50-276-4863 Supplier Grainger Supplier No. 16N602
The Grainger 16N602 is a 1/4 HP, 115V, 60Hz, 1-phase, 3450 RPM, 56C frame, open drip proof, ball bearing, C-face, sleeve bearing electric motor. It has a NEMA 48 frame size with a 1.25" shaft diameter and 1.38" shaft height. The motor is totally enclosed and fan cooled with a Class B insulation and 1.15 service factor. It can output 0.186 kW with a full load amp rating of 5.8 Amps. The 16N602 electric motor is ideal for various light industrial applications.