Learn More
Grainger G3261 Level Gauge HDPE D 101in
Catalog No. 502764205
Encompass_Preferred
The Krueger 101-inch Level Gauge is a liquid level indicator designed for 101-inch containers. It features a clear acrylic tube with gallon graduations and a polypropylene float to provide an accurate visual display of liquid level. This gauge can withstand temperatures up to 180°F and pressures up to 100 PSI. The dimensions are 106 inches x 2 inches diameter. This direct reading gauge is easy to install and requires no power source. It is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-4205 Supplier Grainger Supplier No. 16N364
The Krueger 101-inch Level Gauge is a liquid level indicator designed for 101-inch containers. It features a clear acrylic tube with gallon graduations and a polypropylene float to provide an accurate visual display of liquid level. This gauge can withstand temperatures up to 180°F and pressures up to 100 PSI. The dimensions are 106 inches x 2 inches diameter. This direct reading gauge is easy to install and requires no power source. It is made in the USA by Krueger Sentry Gauge.