Learn More
Grainger G3261 Level Gauge HDPE D 103in
Catalog No. 502764490
Encompass_Preferred
The Grainger website features aluminum and HDPE rod/float liquid level gauges for drums and tanks. These gauges allow visual monitoring of liquid levels, with floats moving up and down rods as liquid levels change. They are available in various sizes and configurations to fit different tank openings and applications. Key features include corrosion resistance, chemical compatibility, easy installation, and economical operation. These gauges provide a simple, reliable way to monitor liquid levels in drums, totes and storage tanks.
Catalog No. 50-276-4490 Supplier Grainger Supplier No. 16N366
The Grainger website features aluminum and HDPE rod/float liquid level gauges for drums and tanks. These gauges allow visual monitoring of liquid levels, with floats moving up and down rods as liquid levels change. They are available in various sizes and configurations to fit different tank openings and applications. Key features include corrosion resistance, chemical compatibility, easy installation, and economical operation. These gauges provide a simple, reliable way to monitor liquid levels in drums, totes and storage tanks.