Learn More
Grainger G3261 Level Gauge HDPE D 106in
Catalog No. 502764558
Encompass_Preferred
The Krueger 106-inch Level Gauge is a liquid level indicator designed for 106-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod as liquid level changes. The gauge has a dual scale that reads from 0-100 inches on one side and 0-250 centimeters on the other. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge helps monitor liquid levels in large tanks and vessels. It has a 3/4 inch NPT bottom connection and comes with PTFE thread sealant.
Catalog No. 50-276-4558 Supplier Grainger Supplier No. 16N369
The Krueger 106-inch Level Gauge is a liquid level indicator designed for 106-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod as liquid level changes. The gauge has a dual scale that reads from 0-100 inches on one side and 0-250 centimeters on the other. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge helps monitor liquid levels in large tanks and vessels. It has a 3/4 inch NPT bottom connection and comes with PTFE thread sealant.