Learn More
Grainger G3261 Level Gauge HDPE D 107in
Catalog No. 502764699
Encompass_Preferred
The Krueger 5MFX1 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and bronze body. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level indicator provides a simple, reliable way to monitor boiler water levels. With no moving parts, it is maintenance free. The 5MFX1 has 3/4 inch NPT connections and is made in the USA.
Catalog No. 50-276-4699 Supplier Grainger Supplier No. 16N370
The Krueger 5MFX1 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and bronze body. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level indicator provides a simple, reliable way to monitor boiler water levels. With no moving parts, it is maintenance free. The 5MFX1 has 3/4 inch NPT connections and is made in the USA.