Learn More
Grainger G3261 Level Gauge HDPE D 18in
Catalog No. 502764509
Encompass_Preferred
The Grainger Approved Level Gauge HDPE is a level gauge made of high-density polyethylene plastic. It has a maximum temperature rating of 140°F and a maximum pressure rating of 150 psi. The gauge has a 2 inch dial size and 1/2 inch NPT male thread connections. It can be used on water, oil and other compatible non-corrosive liquids. The dial cover is clear to allow easy monitoring of liquid level. This economical plastic gauge provides accurate level indication in a wide variety of applications.
Catalog No. 50-276-4509 Supplier Grainger Supplier No. 16N293
The Grainger Approved Level Gauge HDPE is a level gauge made of high-density polyethylene plastic. It has a maximum temperature rating of 140°F and a maximum pressure rating of 150 psi. The gauge has a 2 inch dial size and 1/2 inch NPT male thread connections. It can be used on water, oil and other compatible non-corrosive liquids. The dial cover is clear to allow easy monitoring of liquid level. This economical plastic gauge provides accurate level indication in a wide variety of applications.