Learn More
Grainger G3261 Level Gauge HDPE D 23in
Catalog No. 502764626
Encompass_Preferred
The Krueger 18-Inch Level Gauge is a liquid level indicator designed for 18-inch diameter containers. It features a clear acrylic tube with gallon graduations and a polypropylene float to provide an easy visual reference for liquid level. This gauge can handle temperatures up to 180°F and pressures up to 100 PSI. It has 3/4" NPT male thread connections. The overall length is 22". This economical gauge is ideal for monitoring liquid levels in processing tanks, supply drums, and other industrial containers.
Catalog No. 50-276-4626 Supplier Grainger Supplier No. 16N298
The Krueger 18-Inch Level Gauge is a liquid level indicator designed for 18-inch diameter containers. It features a clear acrylic tube with gallon graduations and a polypropylene float to provide an easy visual reference for liquid level. This gauge can handle temperatures up to 180°F and pressures up to 100 PSI. It has 3/4" NPT male thread connections. The overall length is 22". This economical gauge is ideal for monitoring liquid levels in processing tanks, supply drums, and other industrial containers.