Learn More
Grainger G3261 Level Gauge HDPE D 34in
Catalog No. 502764593
Encompass_Preferred
The Grainger 16N308 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. With its larger striking surface, it can cover more area with each blow. The hammer weighs 1.4 lbs. and is 11 inches long overall. It is made in the USA.
Catalog No. 50-276-4593 Supplier Grainger Supplier No. 16N308
Add to Cart
Add to Cart
The Grainger 16N308 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. With its larger striking surface, it can cover more area with each blow. The hammer weighs 1.4 lbs. and is 11 inches long overall. It is made in the USA.