Learn More
Grainger G3261 Level Gauge HDPE D 37in
Catalog No. 502764496
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include corrosion resistance, chemical compatibility, accuracy within 1/4 inch, and visibility in daylight. They mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. Multiple length options are available to fit different container heights. These affordable, durable gauges provide an easy way to monitor liquid levels.
Catalog No. 50-276-4496 Supplier Grainger Supplier No. 16N310
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include corrosion resistance, chemical compatibility, accuracy within 1/4 inch, and visibility in daylight. They mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. Multiple length options are available to fit different container heights. These affordable, durable gauges provide an easy way to monitor liquid levels.