Learn More
Grainger G3261 Level Gauge HDPE D 39in
Catalog No. 502764457
Encompass_Preferred
The Grainger G3261 Level Gauge is a 39 inch long dipstick made of durable high density polyethylene plastic. It features easy-to-read level markings and a comfortable grip handle. This gauge allows users to conveniently check fluid levels in tanks and reservoirs. With a measurement range of 0 to 39 inches, this device is suitable for a variety of industrial and commercial applications. The HDPE construction ensures chemical resistance and longevity. Overall, the Grainger G3261 is a reliable, heavy-duty dipstick designed for frequent fluid level checks.
Catalog No. 50-276-4457 Supplier Grainger Supplier No. 16N311
The Grainger G3261 Level Gauge is a 39 inch long dipstick made of durable high density polyethylene plastic. It features easy-to-read level markings and a comfortable grip handle. This gauge allows users to conveniently check fluid levels in tanks and reservoirs. With a measurement range of 0 to 39 inches, this device is suitable for a variety of industrial and commercial applications. The HDPE construction ensures chemical resistance and longevity. Overall, the Grainger G3261 is a reliable, heavy-duty dipstick designed for frequent fluid level checks.