Learn More
Grainger G3261 Level Gauge HDPE D 43in
Catalog No. 502763720
Encompass_Preferred
The Grainger 16N314 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is rated for continuous duty applications.
Catalog No. 50-276-3720 Supplier Grainger Supplier No. 16N314
Add to Cart
Add to Cart
The Grainger 16N314 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is rated for continuous duty applications.