Learn More
Grainger G3261 Level Gauge HDPE D 47in
Catalog No. 502764109
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks made of galvanized steel and HDPE. These rod/float gauges allow visual monitoring of liquid levels. They feature corrosion-resistant materials suitable for use with water, chemicals, petroleum products and more. Lengths range from 7" to 78" to fit different size containers. They can be ordered with NPT thread connections in various sizes. These affordable gauges provide reliable level indication in industrial liquid storage tanks.
Catalog No. 50-276-4109 Supplier Grainger Supplier No. 16N316
The Grainger website features liquid level gauges for drums and tanks made of galvanized steel and HDPE. These rod/float gauges allow visual monitoring of liquid levels. They feature corrosion-resistant materials suitable for use with water, chemicals, petroleum products and more. Lengths range from 7" to 78" to fit different size containers. They can be ordered with NPT thread connections in various sizes. These affordable gauges provide reliable level indication in industrial liquid storage tanks.