Learn More
Grainger G3261 Level Gauge HDPE D 50in
Catalog No. 502764459
Encompass_Preferred
The Krueger Level Gauge For 133 in Container is a mechanical level gauge designed for use with 133 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates container level on a clearly marked scale in 1/8 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is made in the USA and enables accurate monitoring of liquid levels.
Catalog No. 50-276-4459 Supplier Grainger Supplier No. 16N318
The Krueger Level Gauge For 133 in Container is a mechanical level gauge designed for use with 133 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates container level on a clearly marked scale in 1/8 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is made in the USA and enables accurate monitoring of liquid levels.