Learn More
Grainger G3261 Level Gauge HDPE D 53in
Catalog No. 502763723
Encompass_Preferred
The Grainger G3261 Level Gauge is a 53 inch long dipstick made of durable high density polyethylene plastic with easy-to-read level markings. It allows users to check fluid levels in tanks and reservoirs. This heavy duty dipstick has a D-shaped handle on top for easy gripping and control. The gauge's measurements are marked in both standard and metric units. Overall, this is a sturdy, no-frills dipstick for reliably checking fluid levels in industrial equipment and machinery.
Catalog No. 50-276-3723 Supplier Grainger Supplier No. 16N321
Add to Cart
Add to Cart
The Grainger G3261 Level Gauge is a 53 inch long dipstick made of durable high density polyethylene plastic with easy-to-read level markings. It allows users to check fluid levels in tanks and reservoirs. This heavy duty dipstick has a D-shaped handle on top for easy gripping and control. The gauge's measurements are marked in both standard and metric units. Overall, this is a sturdy, no-frills dipstick for reliably checking fluid levels in industrial equipment and machinery.