Learn More
Grainger G3261 Level Gauge HDPE D 54in
Catalog No. 502764582
Encompass_Preferred
The Grainger 16N322 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor, thermally protected, manual reset overload protector, and meets NEMA design B standards. This motor can be used for various industrial applications requiring 3/4 HP at 1725 RPM. It requires a 115V power source and measures approximately 9-1/4" long by 7-3/16" wide by 8-7/16" high.
Catalog No. 50-276-4582 Supplier Grainger Supplier No. 16N322
The Grainger 16N322 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor, thermally protected, manual reset overload protector, and meets NEMA design B standards. This motor can be used for various industrial applications requiring 3/4 HP at 1725 RPM. It requires a 115V power source and measures approximately 9-1/4" long by 7-3/16" wide by 8-7/16" high.