Learn More
Grainger G3261 Level Gauge HDPE D 55in
Catalog No. 502763724
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These affordable gauges install easily via existing bung openings and are available in different lengths to fit any size drum or tank. Overall, they are durable, low-maintenance liquid level indicators ideal for industrial applications.
Catalog No. 50-276-3724 Supplier Grainger Supplier No. 16N323
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These affordable gauges install easily via existing bung openings and are available in different lengths to fit any size drum or tank. Overall, they are durable, low-maintenance liquid level indicators ideal for industrial applications.