Learn More
Grainger G3261 Level Gauge HDPE D 56in
Catalog No. 502763725
Encompass_Preferred
The Grainger G3261 Level Gauge is a 56 inch long high-density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/4 inch for precise level indication. The gauge's chemical resistance allows it to be used with water, diesel fuel, antifreeze, hydraulic oil and more. With an operating temperature range of -20°F to 150°F, this durable and economical dipstick is suitable for a variety of industrial and commercial applications. The G3261 helps provide at-a-glance monitoring of liquid levels.
Catalog No. 50-276-3725 Supplier Grainger Supplier No. 16N324
The Grainger G3261 Level Gauge is a 56 inch long high-density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/4 inch for precise level indication. The gauge's chemical resistance allows it to be used with water, diesel fuel, antifreeze, hydraulic oil and more. With an operating temperature range of -20°F to 150°F, this durable and economical dipstick is suitable for a variety of industrial and commercial applications. The G3261 helps provide at-a-glance monitoring of liquid levels.