Learn More
Grainger G3261 Level Gauge HDPE D 58in
Catalog No. 502764052
Encompass_Preferred
The Krueger Level Gauge For 58 in Container is a mechanical level gauge designed for use with 58 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be screwed into a tank opening. This heavy duty mechanical gauge is ideal for use with chemicals, water, petroleum products and other liquids. It provides accurate liquid level indication in a rugged stainless steel construction.
Catalog No. 50-276-4052 Supplier Grainger Supplier No. 16N326
The Krueger Level Gauge For 58 in Container is a mechanical level gauge designed for use with 58 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be screwed into a tank opening. This heavy duty mechanical gauge is ideal for use with chemicals, water, petroleum products and other liquids. It provides accurate liquid level indication in a rugged stainless steel construction.