Learn More
Grainger G3261 Level Gauge HDPE D 59in
Catalog No. 502764468
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges allow you to monitor liquid levels inside containers without having to open them. They are available in rod/float and tape styles, with rod/float gauges using a stainless steel or HDPE float that moves up and down a galvanized steel or HDPE rod as liquid levels change. Tape gauges use a coated steel tape with calibration markings that extends into the tank. Both gauge types allow for easy external monitoring of liquid levels. The gauges are designed for use in a variety of liquids including chemicals, petroleum products, waste water and more. They come in various lengths to fit different sized containers.
Catalog No. 50-276-4468 Supplier Grainger Supplier No. 16N327
The Grainger website features liquid level gauges for drums and tanks. These gauges allow you to monitor liquid levels inside containers without having to open them. They are available in rod/float and tape styles, with rod/float gauges using a stainless steel or HDPE float that moves up and down a galvanized steel or HDPE rod as liquid levels change. Tape gauges use a coated steel tape with calibration markings that extends into the tank. Both gauge types allow for easy external monitoring of liquid levels. The gauges are designed for use in a variety of liquids including chemicals, petroleum products, waste water and more. They come in various lengths to fit different sized containers.