Learn More
Grainger G3261 Level Gauge HDPE D 68in
Catalog No. 502764108
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These gauges mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. They are available in different lengths and materials to suit the application. Overall these are durable, low-maintenance liquid level indicators ideal for industrial use.
Catalog No. 50-276-4108 Supplier Grainger Supplier No. 16N334
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These gauges mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. They are available in different lengths and materials to suit the application. Overall these are durable, low-maintenance liquid level indicators ideal for industrial use.