Learn More
Grainger G3261 Level Gauge HDPE D 69in
Catalog No. 502764532
Encompass_Preferred
The Grainger 16N335 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The hammer has a fiberglass handle that provides durability. This general purpose hammer allows the user to deliver heavy blows without marring the striking surface and is well-suited for assembling equipment or driving machine parts without damage. The hammer weighs approximately 1.6 lbs.
Catalog No. 50-276-4532 Supplier Grainger Supplier No. 16N335
The Grainger 16N335 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The hammer has a fiberglass handle that provides durability. This general purpose hammer allows the user to deliver heavy blows without marring the striking surface and is well-suited for assembling equipment or driving machine parts without damage. The hammer weighs approximately 1.6 lbs.