Learn More
Grainger G3261 Level Gauge HDPE D 70in
Catalog No. 502764594
Encompass_Preferred
The Grainger 16N336 is a 16 oz. aerosol can of CRC Brakleen Brake Parts Cleaner. It is a fast drying, non-flammable brake parts cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. The cleaner sprays out as a liquid and features a 360° valve that allows spraying in any position.
Catalog No. 50-276-4594 Supplier Grainger Supplier No. 16N336
The Grainger 16N336 is a 16 oz. aerosol can of CRC Brakleen Brake Parts Cleaner. It is a fast drying, non-flammable brake parts cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. The cleaner sprays out as a liquid and features a 360° valve that allows spraying in any position.