Learn More
Grainger G3261 Level Gauge HDPE D 73in
Catalog No. 502764595
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without opening the lid. Models include polypropylene and stainless steel construction with clear polycarbonate or acrylic sight tubes. Gauges feature 1" NPT thread connections and are available in 6" to 72" lengths to fit small to large capacity drums and tanks. They can handle temperatures up to 200°F and pressures up to 100 PSI. Overall, these gauges provide a safe, convenient way to monitor liquid levels in industrial containers.
Catalog No. 50-276-4595 Supplier Grainger Supplier No. 16N338
The Grainger website features liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without opening the lid. Models include polypropylene and stainless steel construction with clear polycarbonate or acrylic sight tubes. Gauges feature 1" NPT thread connections and are available in 6" to 72" lengths to fit small to large capacity drums and tanks. They can handle temperatures up to 200°F and pressures up to 100 PSI. Overall, these gauges provide a safe, convenient way to monitor liquid levels in industrial containers.