Learn More
Grainger G3261 Level Gauge HDPE D 76in
Catalog No. 502764524
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges allow you to visually check liquid levels in your containers. They feature either galvanized steel or HDPE float materials. The gauges have threaded process connections to fit standard bung openings. They include either a direct reading scale to show exact liquid height or a colored float that rises with liquid level. These simple mechanical gauges provide an economical way to monitor liquid levels in drums and tanks during filling, storage or dispensing. With no power required, they offer reliable visual monitoring to prevent overfills or dry runs.
Catalog No. 50-276-4524 Supplier Grainger Supplier No. 16N341
The Grainger website features liquid level gauges for drums and tanks. These gauges allow you to visually check liquid levels in your containers. They feature either galvanized steel or HDPE float materials. The gauges have threaded process connections to fit standard bung openings. They include either a direct reading scale to show exact liquid height or a colored float that rises with liquid level. These simple mechanical gauges provide an economical way to monitor liquid levels in drums and tanks during filling, storage or dispensing. With no power required, they offer reliable visual monitoring to prevent overfills or dry runs.