Learn More
Grainger G3261 Level Gauge HDPE D 77in
Catalog No. 502764461
Encompass_Preferred
The Krueger Level Gauge For 134 in Container is a liquid level gauge designed for use with 134 inch diameter containers. It features a stainless steel float and stem with a polypropylene guide tube. The gauge indicates liquid level on a dial face that is 8 inches in diameter. It has a dual scale that reads in inches on the outer ring, and liters on the inner ring. The gauge mounts on top of the container and provides continuous monitoring of liquid level. It has a maximum reading of 80 inches or 2000 liters. This industrial gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-4461 Supplier Grainger Supplier No. 16N342
The Krueger Level Gauge For 134 in Container is a liquid level gauge designed for use with 134 inch diameter containers. It features a stainless steel float and stem with a polypropylene guide tube. The gauge indicates liquid level on a dial face that is 8 inches in diameter. It has a dual scale that reads in inches on the outer ring, and liters on the inner ring. The gauge mounts on top of the container and provides continuous monitoring of liquid level. It has a maximum reading of 80 inches or 2000 liters. This industrial gauge is made in the USA by Krueger Sentry Gauge.